Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ak8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033874.2 | NP_001029046.2 | 11, 12 | 1159 | ||
GenBank mRNA | AB046266.1 | - | 2, 3 | 178 | ||
AK004439.1 | - | 11, 12 | 1157 | |||
BC100481.1 | - | 11, 12 | 1156 | 30917849 | ||
BC109352.2 | - | 11, 12 | 1146 | 40057006 |