Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vmn2r71 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105184.1 | NP_001098654.1 | 3 | 974 |
Gene Symbol: Vmn2r66 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033878.3 | NP_001029050.3 | 3 | 962 | ||
GenBank mRNA | AK156873.1 | - | 1 | 883 |
Set Membership: |
![]() |