Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp5k | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007507.2 | NP_031533.2 | 4 | 279 | ||
GenBank mRNA | AK011342.1 | - | 3 | 189 | ||
BC028438.1 | - | 4 | 279 | 1332765 | ||
BC089550.1 | - | 4 | 271 | 30380642 |