Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Orc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025378.2 | NP_001020549.1 | 10 | 987 | ||
NM_001271526.1 | NP_001258455.1 | 13 | 1512 | |||
NM_008765.3 | NP_032791.1 | 13 | 1518 | |||
GenBank mRNA | AB190256.1 | - | 10 | 977 | ||
AK048408.1 | - | 13 | 1337 | |||
AK051747.1 | - | 13 | 1347 | |||
AK133604.1 | - | 13 | 1338 | |||
AK146281.1 | - | 13 | 1337 | |||
AK146859.1 | - | 13 | 1332 | |||
BC015257.1 | - | 13 | 1328 | 4234871 | ||
U27457.1 | - | 13 | 1337 |