Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pomc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278581.1 | NP_001265510.1 | 4 | 783 | ||
NM_001278582.1 | NP_001265511.1 | 4 | 718 | |||
NM_001278583.1 | NP_001265512.1 | 4 | 688 | |||
NM_001278584.1 | NP_001265513.1 | 3 | 577 | |||
NM_008895.4 | NP_032921.1 | 3 | 607 | |||
GenBank mRNA | AK017492.1 | - | 3 | 603 | ||
AK017581.1 | - | 3 | 572 | |||
AK030714.1 | - | 4 | 682 | |||
AK077426.1 | - | 3 | 571 | |||
AK133592.1 | - | 3 | 602 | |||
AK133646.1 | - | 3 | 602 | |||
AK133740.1 | - | 3 | 601 | |||
AK133776.1 | - | 3 | 603 | |||
AK133800.1 | - | 3 | 603 | |||
BB022229.2 | - | 1 | 263 | |||
BC061215.1 | - | 3 | 601 | 30253829 | ||
M30489.1 | - | 1 | 83 |