Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lgals4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010706.2 | NP_034836.1 | 6 | 924 | ||
GenBank mRNA | AF026795.1 | - | 5 | 474 | ||
AF510729.1 | - | 6 | 573 | |||
AK145734.1 | - | 6 | 744 | |||
AY044870.1 | - | 6 | 596 | |||
BC011236.1 | - | 6 | 562 | 4205669 | ||
BC021632.1 | - | 6 | 580 | 4218531 | ||
BC030297.1 | - | 6 | 737 | 4986324 | ||
BC094008.1 | - | 6 | 575 | 4986911 | ||
BC141106.1 | - | 6 | 646 | 9055718 | ||
BC145387.1 | - | 6 | 646 | 9053945 | ||
EF017938.1 | - | 6 | 924 | |||
EF017939.1 | - | 6 | 924 |