Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr56 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010999.2 | NP_035129.1 | 6 | 1013 | ||
GenBank mRNA | AF102533.1 | - | 1 | 85 | ||
AF102535.1 | - | 1 | 85 | |||
AF102537.1 | - | 1 | 85 | |||
AK053352.1 | - | 6 | 1013 | |||
BC141219.1 | - | 5 | 844 | 9055831 | ||
BC147437.1 | - | 5 | 958 | 9056030 |