Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hs3st3b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018805.2 | NP_061275.2 | 1 | 521 | ||
GenBank mRNA | AF168992.1 | - | 1 | 479 | ||
AK085242.1 | - | 1 | 529 | |||
AK171996.1 | - | 1 | 515 | |||
AK172015.1 | - | 1 | 520 | |||
AK172271.1 | - | 1 | 522 | |||
BC116733.1 | - | 1 | 388 | 40126052 | ||
BC116743.1 | - | 1 | 388 | 40126062 |