Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC12A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134771.1 | NP_001128243.1 | 2 | 267 | ||
NM_020708.4 | NP_065759.1 | 2 | 285 | |||
XM_017027981.1 | XP_016883470.1 | 2 | 267 | |||
GenBank mRNA | AB033002.1 | - | 2 | 100 | ||
AF208159.1 | - | 2 | 122 | |||
AK098371.1 | - | 2 | 271 | |||
AK289758.1 | - | 2 | 244 | |||
AK294059.1 | - | 2 | 214 | |||
AK296331.1 | - | 2 | 243 | |||
BC132668.1 | - | 2 | 210 | 40146690 | ||
BC132670.1 | - | 2 | 210 | 40146692 | ||
DA102113.1 | - | 2 | 267 | |||
DC320473.1 | - | 2 | 286 | |||
KC295693.1 | - | 2 | 285 | |||
KC295694.1 | - | 2 | 285 | |||
KC295695.1 | - | 2 | 285 | |||
KC295698.1 | - | 2 | 285 | |||
KC295699.1 | - | 2 | 285 | |||
KC295700.1 | - | 2 | 285 | |||
KC295701.1 | - | 2 | 285 |