Search Thermo Fisher Scientific
Gene Symbol: Dppa5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025274.3 | NP_079550.2 | 3 | 494 | ||
GenBank mRNA | AF490349.1 | - | 3 | 450 | ||
AK010519.1 | - | 3 | 451 | |||
AK010584.1 | - | 3 | 450 | |||
AK145761.1 | - | 3 | 453 | |||
AV476247.1 | - | 1 | 107 | |||
BC092353.1 | - | 3 | 439 | 30730511 | ||
BC092354.1 | - | 3 | 447 | 30732471 | ||
BC095979.1 | - | 3 | 439 | 30732937 | ||
X57708.1 | - | 3 | 452 |