Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MTUS1 SIRNA Assays ›
Gene Symbol:
MTUS1
Gene Name:
microtubule associated tumor suppressor 1
Gene Aliases:
ATBP, ATIP, ICIS, MP44, MTSG1
Chromosome Location:
Chr.8: 17643794 - 17801579 on Build GRCh38
UniGene ID:
Hs.7946
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
28761
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MTUS1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001001924.2
NP_001001924.1
15
5356
NM_001001925.2
NP_001001925.1
14
5194
NM_001001931.2
NP_001001931.1
12
2943
NM_001166393.1
NP_001159865.1
12
2588
NM_020749.4
NP_065800.1
10
2740
XM_005273577.3
XP_005273634.1
15
5264
XM_005273578.3
XP_005273635.1
15
5352
XM_005273579.3
XP_005273636.1
15
5268
XM_005273580.3
XP_005273637.1
14
5102
XM_005273581.3
XP_005273638.1
14
5190
XM_005273582.3
XP_005273639.1
14
5106
XM_011544595.2
XP_011542897.1
15
5298
XM_017013698.1
XP_016869187.1
16
5270
XM_017013699.1
XP_016869188.1
16
5492
XM_017013700.1
XP_016869189.1
15
5155
XM_017013701.1
XP_016869190.1
16
5272
XM_017013702.1
XP_016869191.1
14
4927
XM_017013704.1
XP_016869193.1
9
2262
GenBank mRNA
AB033114.1
-
14
4804
AF121259.1
-
10
2376
AF394227.1
-
1
1073
AK000172.1
-
3
1477
AK024357.1
-
9
2262
AK026661.1
-
1
848
AK125188.1
-
12
2939
AL096842.1
-
15
4144
BC033842.1
-
7
1828
BC136320.1
-
15
5103
9020307
BC142971.1
-
10
2669
BX648879.1
-
8
3242
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-qqtbl:80/10.239.164.61:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0