Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ms4a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025610.4 | NP_001020781.1 | 3 | 317 | ||
NM_001276398.1 | NP_001263327.1 | 3 | 557 | |||
NM_027836.6 | NP_082112.3 | 3 | 317 | |||
GenBank mRNA | AF237917.2 | - | 3 | 268 | ||
AK018686.1 | - | 3 | 288 | |||
AK046559.1 | - | 3 | 318 | |||
AK079888.1 | - | 3 | 558 | |||
AK085297.1 | - | 3 | 253 | |||
AK149890.1 | - | 3 | 285 | |||
AK151261.1 | - | 3 | 288 | |||
AK152098.1 | - | 3 | 288 | |||
BC019784.1 | - | 3 | 264 | |||
BC024402.1 | - | 3 | 276 | 5035752 |