Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slamf8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029084.3 | NP_083360.2 | 2, 3 | 498 | ||
GenBank mRNA | AB196828.1 | - | 2, 3 | 362 | ||
AB196829.1 | - | 2, 3 | 362 | |||
AB196830.1 | - | 2, 3 | 362 | |||
AB196831.1 | - | 2, 3 | 362 | |||
AK017911.1 | - | 2, 3 | 401 | |||
AK142633.1 | - | 2, 3 | 499 | |||
AK156077.1 | - | 2, 3 | 397 | |||
BC024587.1 | - | 2, 3 | 418 | 5066283 |