Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Yif1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110201.1 | NP_001103671.1 | 7 | 774 | ||
NM_029887.3 | NP_084163.2 | 7 | 783 | |||
GenBank mRNA | AK020444.1 | - | 7 | 754 | ||
AK135529.1 | - | 752 | ||||
BC109331.2 | - | 7 | 753 | 40051521 |