Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FAM135A SIRNA Assays ›
Gene Symbol:
FAM135A
Gene Name:
family with sequence similarity 135 member A
Gene Aliases:
KIAA1411
Chromosome Location:
Chr.6: 70413404 - 70561174 on Build GRCh38
UniGene ID:
Hs.211700
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
28778
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FAM135A
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001105531.2
NP_001099001.1
8
965
NM_001162529.1
NP_001156001.1
6
580
NM_020819.4
NP_065870.3
6
750
XM_005248743.1
XP_005248800.1
8
883
XM_005248744.4
XP_005248801.1
7
850
XM_005248745.2
XP_005248802.1
8
883
XM_005248748.2
XP_005248805.1
8
883
XM_005248749.1
XP_005248806.1
8
883
XM_005248751.2
XP_005248808.1
8
883
XM_011535993.2
XP_011534295.1
7
603
XM_011535994.2
XP_011534296.1
8
1023
XM_011535995.2
XP_011534297.1
8
824
XM_011535996.2
XP_011534298.1
8
981
XM_011535997.2
XP_011534299.1
6
725
XM_017011124.1
XP_016866613.1
7
856
XM_017011125.1
XP_016866614.1
7
706
XM_017011126.1
XP_016866615.1
8
1020
XM_017011127.1
XP_016866616.1
7
802
XM_017011128.1
XP_016866617.1
6
726
XM_017011129.1
XP_016866618.1
8
1039
XM_017011130.1
XP_016866619.1
7
764
XM_017011131.1
XP_016866620.1
8
865
XM_017011132.1
XP_016866621.1
7
852
XM_017011133.1
XP_016866622.1
6
642
XM_017011134.1
XP_016866623.1
8
883
XM_017011135.1
XP_016866624.1
8
937
GenBank mRNA
AB037832.1
-
6
548
AK000183.1
-
5
415
AK225230.1
-
5
415
BC131782.1
-
8
791
40124841
BM461890.1
-
3
263
CR749254.1
-
6
588
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
cellular lipid metabolic process
carboxylic ester hydrolase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-46vz9:80/10.239.163.144:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0