Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr847 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146525.1 | NP_666736.1 | 1 | 929 | ||
GenBank mRNA | BC111580.1 | - | 1 | 928 |