Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr76 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146682.1 | NP_666893.1 | 1 | 643 | ||
GenBank mRNA | BC138072.1 | - | 1 | 669 | 8861091 | |
BC138073.1 | - | 1 | 669 | 8861092 | ||
Y15525.1 | - | 1 | 457 |