Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sacs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172809.3 | NP_766397.2 | 8 | 2369 | ||
GenBank mRNA | AK038655.1 | - | 2 | 1933 | ||
AK046501.1 | - | 7 | 2224 | |||
AK053626.1 | - | 7 | 1804 | |||
AK160797.1 | - | 8 | 2370 | |||
BC138482.1 | - | 7 | 1925 | 8861502 | ||
BC138483.1 | - | 7 | 1925 | 8861503 | ||
BC171956.1 | - | 7 | 1925 | 9054242 |