Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alkbh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175016.2 | NP_778181.2 | 3, 4 | 752 | ||
GenBank mRNA | AK079195.1 | - | 3, 4 | 589 | ||
BC062188.1 | - | 3, 4 | 752 | 30146542 |