Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Txndc15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175150.3 | NP_780359.2 | 4 | 896 | ||
GenBank mRNA | AK009878.1 | - | 4 | 822 | ||
AK079517.1 | - | 4 | 843 | |||
AK160051.1 | - | 4 | 865 | |||
AK167701.1 | - | 4 | 804 | |||
AK167838.1 | - | 4 | 862 | |||
AK167977.1 | - | 4 | 861 | |||
BC029015.1 | - | 3 | 632 | |||
BC038082.1 | - | 2 | 154 | |||
BC062180.1 | - | 4 | 896 | 6336176 | ||
BC094275.1 | - | 4 | 732 |