Gene Symbol: Txnl4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038608.2 | NP_001033697.2 | 4 | 976 | ||
NM_001042408.1 | NP_001035867.1 | 4 | 952 | |||
NM_178604.3 | NP_848719.2 | 4 | 1088 | |||
GenBank mRNA | AK010599.1 | - | 4 | 976 | ||
AK011717.1 | - | 4 | 1040 | |||
AK171121.1 | - | 4 | 928 | |||
BC003876.1 | - | 4 | 904 | |||
BC014770.1 | - | 4 | 952 | |||
BC025053.1 | - | 4 | 929 | |||
BC029327.1 | - | 4 | 946 |