Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCUBE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170690.1 | NP_001164161.1 | 1, 2 | 212 | ||
NM_020974.2 | NP_066025.2 | 1, 2 | 212 | |||
XM_005253032.4 | XP_005253089.2 | 1, 2 | 208 | |||
XM_005253033.4 | XP_005253090.2 | 1, 2 | 208 | |||
XM_005253034.4 | XP_005253091.2 | 1, 2 | 208 | |||
XM_005253035.4 | XP_005253092.2 | 1, 2 | 208 | |||
XM_005253036.4 | XP_005253093.2 | 1, 2 | 208 | |||
XM_005253037.4 | XP_005253094.2 | 1, 2 | 208 | |||
XM_011520246.2 | XP_011518548.1 | 1, 2 | 208 | |||
XM_011520247.2 | XP_011518549.1 | 1, 2 | 208 | |||
XM_011520248.2 | XP_011518550.1 | 1, 2 | 208 | |||
XM_017018080.1 | XP_016873569.1 | 1, 2 | 208 | |||
XM_017018081.1 | XP_016873570.1 | 1, 2 | 208 | |||
XM_017018082.1 | XP_016873571.1 | 1, 2 | 208 | |||
GenBank mRNA | AK123039.1 | - | 1, 2 | 208 | ||
AK131552.1 | - | 1, 2 | 206 | |||
BC111690.1 | - | 1, 2 | 155 | 40008764 | ||
DN995815.1 | - | 1, 2 | 240 |