Search Thermo Fisher Scientific
Gene Symbol: B3GALT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020981.3 | NP_066191.1 | 2 | 493 | ||
XM_005246931.3 | XP_005246988.1 | 5 | 927 | |||
XM_006712819.3 | XP_006712882.1 | 3 | 5201 | |||
XM_011512085.2 | XP_011510387.1 | 5 | 943 | |||
GenBank mRNA | AF117222.1 | - | 1 | 234 | ||
BC101545.1 | - | 1 | 193 | 8069051 | ||
BC104813.1 | - | 1 | 193 | 8143816 | ||
E07739.1 | - | 5 | 817 |