Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: 1190005I06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_197988.1 | NP_932105.2 | 3, 4 | 309 | ||
GenBank mRNA | AK004494.1 | - | 3, 4 | 258 | ||
BC027666.1 | - | 3, 4 | 309 | 1245153 |
Gene Symbol: Emc8-1190005i06rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BQ746342.1 | - | 3, 4 | 457 | ||
CX224265.1 | - | 3, 4 | 520 |