Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Raet1e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198193.2 | NP_937836.1 | 4 | 373 | ||
GenBank mRNA | AK012289.1 | - | 4 | 373 | ||
AK131854.1 | - | 6 | 676 | |||
AY054973.1 | - | 2 | 239 | |||
AY056835.1 | - | 2 | 239 | |||
BC132308.1 | - | 2 | 276 | 40130585 | ||
FJ594067.1 | - | 5 | 477 | |||
FJ594068.1 | - | 4 | 407 |