Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfatc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164109.1 | NP_001157581.1 | 8 | 2383 | ||
NM_001164110.1 | NP_001157582.1 | 8 | 2383 | |||
NM_001164111.1 | NP_001157583.1 | 8 | 2091 | |||
NM_001164112.1 | NP_001157584.1 | 8 | 2091 | |||
NM_016791.4 | NP_058071.2 | 8 | 2091 | |||
NM_198429.2 | NP_940821.1 | 8 | 2383 | |||
GenBank mRNA | AF049606.1 | - | 8 | 2044 | ||
AF087434.1 | - | 8 | 2078 | |||
AF239169.1 | - | 8 | 2081 | |||
AK004810.1 | - | 8 | 2049 | |||
AK048610.1 | - | 8 | 2385 | |||
BC061509.1 | - | 8 | 2370 | 5354603 | ||
EU887567.1 | - | 8 | 2078 | |||
EU887568.1 | - | 8 | 2078 | |||
EU887569.1 | - | 8 | 2036 | |||
EU887570.1 | - | 8 | 2078 | |||
EU887571.1 | - | 8 | 2036 | |||
EU887572.1 | - | 8 | 2036 |