Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dlx5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010056.3 | NP_034186.2 | 3 | 1307 | ||
NM_198854.2 | NP_942151.1 | 3 | 1225 | |||
GenBank mRNA | AF033011.1 | - | 3 | 1147 | ||
AF072452.1 | - | 3 | 1307 | |||
AF072453.1 | - | 3 | 1225 | |||
AK134920.1 | - | 3 | 1183 | |||
AK158772.1 | - | 3 | 1242 | |||
U67840.1 | - | 4 | 1283 |