Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ugt1a6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201410.3 | NP_958812.3 | 2 | 469 | ||
GenBank mRNA | AY227198.1 | - | 1 | 425 | ||
BC026561.1 | - | 2 | 449 | 5050488 |