Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFHR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166624.1 | NP_001160096.1 | 5 | 762 | ||
NM_021023.5 | NP_066303.2 | 6 | 945 | |||
GenBank mRNA | AK298459.1 | - | 5 | 746 | ||
BC058009.1 | - | 6 | 903 | 4716505 | ||
X68679.1 | - | 6 | 884 |