Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004720.2 | NP_001004720.1 | 4 | 2106 | ||
NM_001004722.3 | NP_001004722.1 | 4 | 1494 | |||
NM_003581.4 | NP_003572.2 | 5 | 2216 | |||
XM_006712797.3 | XP_006712860.1 | 5 | 2872 | |||
XM_011511991.2 | XP_011510293.1 | 5 | 2310 | |||
XM_011511992.2 | XP_011510294.2 | 3 | 1438 | |||
XM_017005103.1 | XP_016860592.1 | 6 | 2474 | |||
XM_017005104.1 | XP_016860593.1 | 5 | 2486 | |||
XM_017005105.1 | XP_016860594.1 | 5 | 2223 | |||
GenBank mRNA | BC000103.2 | - | 4 | 1994 | 3505427 | |
BC007195.1 | - | 4 | 2001 | 3608149 |