Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SORBS1 SIRNA Assays ›
Gene Symbol:
SORBS1
Gene Name:
sorbin and SH3 domain containing 1
Gene Aliases:
CAP, FLAF2, R85FL, SH3D5, SH3P12, SORB1
Chromosome Location:
Chr.10: 95311773 - 95561420 on Build GRCh38
UniGene ID:
Hs.38621
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
288594
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SORBS1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001034954.1
NP_001030126.1
27
2834
NM_001034955.1
NP_001030127.1
27
2900
NM_001034956.1
NP_001030128.1
24
2387
NM_001034957.1
NP_001030129.1
19
1952
NM_001290294.1
NP_001277223.1
27
2883
NM_001290295.1
NP_001277224.1
22
2166
NM_001290296.1
NP_001277225.1
25
2744
NM_001290297.1
NP_001277226.1
22
2165
NM_001290298.1
NP_001277227.1
23
2270
NM_006434.2
NP_006425.2
18
1784
NM_015385.3
NP_056200.1
22
2367
NM_024991.2
NP_079267.1
23
2262
XM_006717589.1
XP_006717652.1
27
2884
XM_006717593.1
XP_006717656.1
21
2117
XM_011539140.1
XP_011537442.1
29
3628
XM_011539150.1
XP_011537452.1
28
3553
XM_011539155.1
XP_011537457.1
23
3203
XM_011539167.1
XP_011537469.1
22
2486
XM_017015500.1
XP_016870989.1
24
3133
XM_017015501.1
XP_016870990.1
28
3553
XM_017015502.1
XP_016870991.1
27
3460
XM_017015503.1
XP_016870992.1
26
3385
XM_017015504.1
XP_016870993.1
27
2542
XM_017015505.1
XP_016870994.1
25
3301
XM_017015506.1
XP_016870995.1
26
2467
XM_017015507.1
XP_016870996.1
26
3292
XM_017015508.1
XP_016870997.1
25
2374
XM_017015509.1
XP_016870998.1
25
2396
XM_017015510.1
XP_016870999.1
24
3133
XM_017015511.1
XP_016871000.1
24
2290
XM_017015512.1
XP_016871001.1
22
2201
XM_017015513.1
XP_016871002.1
22
2192
XM_017015514.1
XP_016871003.1
22
2108
XM_017015515.1
XP_016871004.1
26
2743
XM_017015516.1
XP_016871005.1
25
2716
XM_017015517.1
XP_016871006.1
25
2659
XM_017015518.1
XP_016871007.1
25
2672
XM_017015519.1
XP_016871008.1
24
2645
XM_017015520.1
XP_016871009.1
24
2584
XM_017015521.1
XP_016871010.1
24
2597
XM_017015522.1
XP_016871011.1
23
2557
XM_017015523.1
XP_016871012.1
23
2513
XM_017015524.1
XP_016871013.1
22
2486
XM_017015525.1
XP_016871014.1
26
2471
XM_017015526.1
XP_016871015.1
25
2374
XM_017015527.1
XP_016871016.1
25
2396
XM_017015528.1
XP_016871017.1
24
2347
XM_017015529.1
XP_016871018.1
24
2299
XM_017015530.1
XP_016871019.1
24
2290
XM_017015531.1
XP_016871020.1
24
2303
XM_017015532.1
XP_016871021.1
23
2272
XM_017015533.1
XP_016871022.1
23
2276
XM_017015534.1
XP_016871023.1
24
2206
XM_017015535.1
XP_016871024.1
23
2228
XM_017015536.1
XP_016871025.1
23
2219
XM_017015537.1
XP_016871026.1
22
2188
XM_017015538.1
XP_016871027.1
23
2179
XM_017015539.1
XP_016871028.1
22
2201
XM_017015540.1
XP_016871029.1
22
2192
GenBank mRNA
AB037717.1
-
23
2194
AF136380.1
-
17
1739
AF136381.1
-
24
2699
AF330623.1
-
21
2120
AF330624.1
-
7
818
AF356525.1
-
27
2845
AF356526.1
-
27
2911
AF356527.1
-
24
2398
AJ489942.1
-
22
2225
AK294924.1
-
11
1144
AK300407.1
-
22
2119
AK304130.1
-
11
1207
AL117472.1
-
22
2325
AM260536.1
-
27
2841
BC042612.1
-
22
2666
BC152463.1
-
23
2194
8862599
BX640814.1
-
11
2197
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
muscle contraction
cell-matrix adhesion
insulin receptor signaling pathway
positive regulation of signal transduction
glucose transport
cellular response to insulin stimulus
stress fiber assembly
positive regulation of glycogen biosynthetic process
positive regulation of glucose import
positive regulation of lipid biosynthetic process
focal adhesion assembly
positive regulation of establishment of protein localization to plasma membrane
actin binding
SH3/SH2 adaptor activity
insulin receptor binding
protein binding
cytoskeletal protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-d4wl8:80/10.239.162.132:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0