Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SORBS1 SIRNA Assays ›
Gene Symbol:
SORBS1
Gene Name:
sorbin and SH3 domain containing 1
Gene Aliases:
CAP, FLAF2, R85FL, SH3D5, SH3P12, SORB1
Chromosome Location:
Chr.10: 95311773 - 95561420 on Build GRCh38
UniGene ID:
Hs.38621
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
288598
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SORBS1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001034954.1
NP_001030126.1
31
7080
NM_001034955.1
NP_001030127.1
31
6657
NM_001034956.1
NP_001030128.1
28
5919
NM_001034957.1
NP_001030129.1
22
5424
NM_001290294.1
NP_001277223.1
32
7189
NM_001290295.1
NP_001277224.1
25
5638
NM_001290296.1
NP_001277225.1
28
6216
NM_001290297.1
NP_001277226.1
25
5637
NM_001290298.1
NP_001277227.1
26
5742
NM_006434.2
NP_006425.2
21
5256
NM_015385.3
NP_056200.1
25
5839
NM_024991.2
NP_079267.1
26
5734
XM_006717589.1
XP_006717652.1
30
6356
XM_006717593.1
XP_006717656.1
24
5589
XM_011539140.1
XP_011537442.1
34
7934
XM_011539150.1
XP_011537452.1
33
7859
XM_011539155.1
XP_011537457.1
28
7509
XM_011539167.1
XP_011537469.1
27
6792
XM_017015500.1
XP_016870989.1
29
7439
XM_017015501.1
XP_016870990.1
31
7025
XM_017015502.1
XP_016870991.1
30
6932
XM_017015503.1
XP_016870992.1
29
6857
XM_017015504.1
XP_016870993.1
32
6848
XM_017015505.1
XP_016870994.1
28
6773
XM_017015506.1
XP_016870995.1
31
6773
XM_017015507.1
XP_016870996.1
29
6764
XM_017015508.1
XP_016870997.1
30
6680
XM_017015509.1
XP_016870998.1
30
6702
XM_017015510.1
XP_016870999.1
27
6605
XM_017015511.1
XP_016871000.1
29
6596
XM_017015512.1
XP_016871001.1
27
6507
XM_017015513.1
XP_016871002.1
27
6498
XM_017015514.1
XP_016871003.1
27
6414
XM_017015515.1
XP_016871004.1
29
6215
XM_017015516.1
XP_016871005.1
28
6188
XM_017015517.1
XP_016871006.1
28
6131
XM_017015518.1
XP_016871007.1
28
6144
XM_017015519.1
XP_016871008.1
27
6117
XM_017015520.1
XP_016871009.1
27
6056
XM_017015521.1
XP_016871010.1
27
6069
XM_017015522.1
XP_016871011.1
26
6029
XM_017015523.1
XP_016871012.1
26
5985
XM_017015524.1
XP_016871013.1
25
5958
XM_017015525.1
XP_016871014.1
29
5943
XM_017015526.1
XP_016871015.1
28
5846
XM_017015527.1
XP_016871016.1
28
5868
XM_017015528.1
XP_016871017.1
27
5819
XM_017015529.1
XP_016871018.1
27
5771
XM_017015530.1
XP_016871019.1
27
5762
XM_017015531.1
XP_016871020.1
27
5775
XM_017015532.1
XP_016871021.1
26
5744
XM_017015533.1
XP_016871022.1
26
5748
XM_017015534.1
XP_016871023.1
27
5678
XM_017015535.1
XP_016871024.1
26
5700
XM_017015536.1
XP_016871025.1
26
5691
XM_017015537.1
XP_016871026.1
25
5660
XM_017015538.1
XP_016871027.1
26
5651
XM_017015539.1
XP_016871028.1
25
5673
XM_017015540.1
XP_016871029.1
25
5664
GenBank mRNA
AB037717.1
-
26
5667
AL117472.1
-
25
5797
AL122082.1
-
1
2498
AL832353.1
-
6
4260
AM260536.1
-
32
7148
BC152463.1
-
26
5667
8862599
BX640814.1
-
15
5729
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
muscle contraction
cell-matrix adhesion
insulin receptor signaling pathway
positive regulation of signal transduction
glucose transport
cellular response to insulin stimulus
stress fiber assembly
positive regulation of glycogen biosynthetic process
positive regulation of glucose import
positive regulation of lipid biosynthetic process
focal adhesion assembly
positive regulation of establishment of protein localization to plasma membrane
actin binding
SH3/SH2 adaptor activity
insulin receptor binding
protein binding
cytoskeletal protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-d4wl8:80/10.239.162.132:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0