Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SORBS1 SIRNA Assays ›
Gene Symbol:
SORBS1
Gene Name:
sorbin and SH3 domain containing 1
Gene Aliases:
CAP, FLAF2, R85FL, SH3D5, SH3P12, SORB1
Chromosome Location:
Chr.10: 95311773 - 95561420 on Build GRCh38
UniGene ID:
Hs.38621
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
288599
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SORBS1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001034954.1
NP_001030126.1
31
4461
NM_001034955.1
NP_001030127.1
31
4038
NM_001034956.1
NP_001030128.1
28
3300
NM_001034957.1
NP_001030129.1
22
2805
NM_001290294.1
NP_001277223.1
32
4570
NM_001290295.1
NP_001277224.1
25
3019
NM_001290296.1
NP_001277225.1
28
3597
NM_001290297.1
NP_001277226.1
25
3018
NM_001290298.1
NP_001277227.1
26
3123
NM_006434.2
NP_006425.2
21
2637
NM_015385.3
NP_056200.1
25
3220
NM_024991.2
NP_079267.1
26
3115
XM_006717589.1
XP_006717652.1
30
3737
XM_006717593.1
XP_006717656.1
24
2970
XM_011539140.1
XP_011537442.1
34
5315
XM_011539150.1
XP_011537452.1
33
5240
XM_011539155.1
XP_011537457.1
28
4890
XM_011539167.1
XP_011537469.1
27
4173
XM_017015500.1
XP_016870989.1
29
4820
XM_017015501.1
XP_016870990.1
31
4406
XM_017015502.1
XP_016870991.1
30
4313
XM_017015503.1
XP_016870992.1
29
4238
XM_017015504.1
XP_016870993.1
32
4229
XM_017015505.1
XP_016870994.1
28
4154
XM_017015506.1
XP_016870995.1
31
4154
XM_017015507.1
XP_016870996.1
29
4145
XM_017015508.1
XP_016870997.1
30
4061
XM_017015509.1
XP_016870998.1
30
4083
XM_017015510.1
XP_016870999.1
27
3986
XM_017015511.1
XP_016871000.1
29
3977
XM_017015512.1
XP_016871001.1
27
3888
XM_017015513.1
XP_016871002.1
27
3879
XM_017015514.1
XP_016871003.1
27
3795
XM_017015515.1
XP_016871004.1
29
3596
XM_017015516.1
XP_016871005.1
28
3569
XM_017015517.1
XP_016871006.1
28
3512
XM_017015518.1
XP_016871007.1
28
3525
XM_017015519.1
XP_016871008.1
27
3498
XM_017015520.1
XP_016871009.1
27
3437
XM_017015521.1
XP_016871010.1
27
3450
XM_017015522.1
XP_016871011.1
26
3410
XM_017015523.1
XP_016871012.1
26
3366
XM_017015524.1
XP_016871013.1
25
3339
XM_017015525.1
XP_016871014.1
29
3324
XM_017015526.1
XP_016871015.1
28
3227
XM_017015527.1
XP_016871016.1
28
3249
XM_017015528.1
XP_016871017.1
27
3200
XM_017015529.1
XP_016871018.1
27
3152
XM_017015530.1
XP_016871019.1
27
3143
XM_017015531.1
XP_016871020.1
27
3156
XM_017015532.1
XP_016871021.1
26
3125
XM_017015533.1
XP_016871022.1
26
3129
XM_017015534.1
XP_016871023.1
27
3059
XM_017015535.1
XP_016871024.1
26
3081
XM_017015536.1
XP_016871025.1
26
3072
XM_017015537.1
XP_016871026.1
25
3041
XM_017015538.1
XP_016871027.1
26
3032
XM_017015539.1
XP_016871028.1
25
3054
XM_017015540.1
XP_016871029.1
25
3045
GenBank mRNA
AB037717.1
-
26
3047
AL117472.1
-
25
3178
AL832353.1
-
6
1645
AM260536.1
-
32
4529
BC152463.1
-
26
3047
8862599
BX640814.1
-
15
3110
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
muscle contraction
cell-matrix adhesion
insulin receptor signaling pathway
positive regulation of signal transduction
glucose transport
cellular response to insulin stimulus
stress fiber assembly
positive regulation of glycogen biosynthetic process
positive regulation of glucose import
positive regulation of lipid biosynthetic process
focal adhesion assembly
positive regulation of establishment of protein localization to plasma membrane
actin binding
SH3/SH2 adaptor activity
insulin receptor binding
protein binding
cytoskeletal protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-8gqds:80/10.239.164.183:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0