Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SORBS1 SIRNA Assays ›
Gene Symbol:
SORBS1
Gene Name:
sorbin and SH3 domain containing 1
Gene Aliases:
CAP, FLAF2, R85FL, SH3D5, SH3P12, SORB1
Chromosome Location:
Chr.10: 95311773 - 95561420 on Build GRCh38
UniGene ID:
Hs.38621
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
288601
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SORBS1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001034954.1
NP_001030126.1
20
1972
NM_001034955.1
NP_001030127.1
20
2038
NM_001034956.1
NP_001030128.1
17
1525
NM_001034957.1
NP_001030129.1
12
1090
NM_001290294.1
NP_001277223.1
20
2021
NM_001290295.1
NP_001277224.1
15
1304
NM_001290296.1
NP_001277225.1
18
1882
NM_001290297.1
NP_001277226.1
15
1303
NM_001290298.1
NP_001277227.1
16
1408
NM_006434.2
NP_006425.2
12
1090
NM_015385.3
NP_056200.1
16
1673
NM_024991.2
NP_079267.1
16
1400
XM_006717589.1
XP_006717652.1
20
2022
XM_006717593.1
XP_006717656.1
14
1255
XM_011539140.1
XP_011537442.1
22
2766
XM_011539150.1
XP_011537452.1
21
2691
XM_011539155.1
XP_011537457.1
16
2341
XM_011539167.1
XP_011537469.1
15
1624
XM_017015500.1
XP_016870989.1
18
2439
XM_017015501.1
XP_016870990.1
21
2691
XM_017015502.1
XP_016870991.1
20
2598
XM_017015503.1
XP_016870992.1
19
2523
XM_017015504.1
XP_016870993.1
20
1680
XM_017015505.1
XP_016870994.1
18
2439
XM_017015506.1
XP_016870995.1
19
1605
XM_017015507.1
XP_016870996.1
20
2598
XM_017015508.1
XP_016870997.1
18
1512
XM_017015509.1
XP_016870998.1
18
1534
XM_017015510.1
XP_016870999.1
18
2439
XM_017015511.1
XP_016871000.1
17
1428
XM_017015512.1
XP_016871001.1
15
1339
XM_017015513.1
XP_016871002.1
15
1330
XM_017015514.1
XP_016871003.1
16
1414
XM_017015515.1
XP_016871004.1
19
1881
XM_017015516.1
XP_016871005.1
18
1854
XM_017015517.1
XP_016871006.1
18
1797
XM_017015518.1
XP_016871007.1
18
1810
XM_017015519.1
XP_016871008.1
17
1783
XM_017015520.1
XP_016871009.1
17
1722
XM_017015521.1
XP_016871010.1
17
1735
XM_017015522.1
XP_016871011.1
16
1695
XM_017015523.1
XP_016871012.1
16
1651
XM_017015524.1
XP_016871013.1
15
1624
XM_017015525.1
XP_016871014.1
19
1609
XM_017015526.1
XP_016871015.1
18
1512
XM_017015527.1
XP_016871016.1
18
1534
XM_017015528.1
XP_016871017.1
17
1485
XM_017015529.1
XP_016871018.1
17
1437
XM_017015530.1
XP_016871019.1
17
1428
XM_017015531.1
XP_016871020.1
17
1441
XM_017015532.1
XP_016871021.1
16
1410
XM_017015533.1
XP_016871022.1
16
1414
XM_017015534.1
XP_016871023.1
18
1512
XM_017015535.1
XP_016871024.1
16
1366
XM_017015536.1
XP_016871025.1
16
1357
XM_017015537.1
XP_016871026.1
15
1326
XM_017015538.1
XP_016871027.1
17
1485
XM_017015539.1
XP_016871028.1
15
1339
XM_017015540.1
XP_016871029.1
15
1330
GenBank mRNA
AB037717.1
-
16
1332
AF330623.1
-
14
1258
AF356525.1
-
20
1983
AF356526.1
-
20
2049
AF356527.1
-
17
1536
AJ489942.1
-
15
1363
AK294924.1
-
4
282
AK300407.1
-
15
1257
AK304130.1
-
4
345
AK304753.1
-
16
2316
AL117472.1
-
16
1631
AM260536.1
-
20
1979
BC042612.1
-
14
1697
BC152463.1
-
16
1332
8862599
BX640814.1
-
4
1335
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
muscle contraction
cell-matrix adhesion
insulin receptor signaling pathway
positive regulation of signal transduction
glucose transport
cellular response to insulin stimulus
stress fiber assembly
positive regulation of glycogen biosynthetic process
positive regulation of glucose import
positive regulation of lipid biosynthetic process
focal adhesion assembly
positive regulation of establishment of protein localization to plasma membrane
actin binding
SH3/SH2 adaptor activity
insulin receptor binding
protein binding
cytoskeletal protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-xql29:80/10.239.165.153:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0