Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYLK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321309.1 | NP_001308238.1 | 33 | 5976 | ||
NM_053025.3 | NP_444253.3 | 34 | 6184 | |||
NM_053026.3 | NP_444254.3 | 33 | 5977 | |||
NM_053027.3 | NP_444255.3 | 33 | 6031 | |||
NM_053028.3 | NP_444256.3 | 32 | 5824 | |||
XM_017006469.1 | XP_016861958.1 | 17 | 3460 | |||
XM_017006470.1 | XP_016861959.1 | 16 | 2448 | |||
XM_017006471.1 | XP_016861960.1 | 16 | 2501 | |||
XM_017006472.1 | XP_016861961.1 | 4 | 794 | |||
XM_017006473.1 | XP_016861962.1 | 4 | 743 | |||
GenBank mRNA | AF172084.1 | - | 1 | 48 | ||
AK127437.1 | - | 1 | 1706 | |||
BC064420.1 | - | 3 | 857 | |||
BC113456.1 | - | 3 | 664 | |||
BC113458.1 | - | 3 | 660 | |||
BM723621.1 | - | 1 | 145 | |||
CR749364.1 | - | 3 | 5643 |