Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PIK3R1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242466.1 | NP_001229395.1 | 9 | 4103 | ||
NM_181504.3 | NP_852556.2 | 10 | 4240 | |||
NM_181523.2 | NP_852664.1 | 16 | 5540 | |||
NM_181524.1 | NP_852665.1 | 10 | 4372 | |||
XM_005248542.3 | XP_005248599.1 | 16 | 5563 | |||
XM_011543493.2 | XP_011541795.1 | 15 | 5193 | |||
XM_017009585.1 | XP_016865074.1 | 16 | 6332 | |||
XM_017009586.1 | XP_016865075.1 | 15 | 5153 | |||
GenBank mRNA | BE888150.1 | - | 1 | 255 | ||
BQ723333.1 | - | 1 | 698 |