Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIK3R1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242466.1 | NP_001229395.1 | 3 | 369 | ||
NM_181504.3 | NP_852556.2 | 4 | 506 | |||
NM_181523.2 | NP_852664.1 | 10 | 1806 | |||
NM_181524.1 | NP_852665.1 | 4 | 638 | |||
XM_005248542.3 | XP_005248599.1 | 10 | 1829 | |||
XM_011543493.2 | XP_011541795.1 | 9 | 1459 | |||
XM_017009585.1 | XP_016865074.1 | 10 | 2598 | |||
XM_017009586.1 | XP_016865075.1 | 9 | 1419 | |||
GenBank mRNA | AF279367.1 | - | 4 | 638 | ||
AK000121.1 | - | 4 | 552 | |||
AK094785.1 | - | 3 | 369 | |||
AK126345.1 | - | 3 | 1769 | |||
AK223613.1 | - | 4 | 486 | |||
AK294919.1 | - | 2 | 625 | |||
AK299743.1 | - | 8 | 835 | |||
BC030815.1 | - | 4 | 478 | 4290954 | ||
BC094795.1 | - | 10 | 1786 | 30528412 | ||
DA487709.1 | - | 4 | 506 | |||
JX133164.1 | - | 3 | 228 | |||
JX133165.1 | - | 3 | 228 | |||
M61906.1 | - | 9 | 1268 |