Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PIK3R1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242466.1 | NP_001229395.1 | 9 | 1497 | ||
NM_181504.3 | NP_852556.2 | 10 | 1634 | |||
NM_181523.2 | NP_852664.1 | 16 | 2934 | |||
NM_181524.1 | NP_852665.1 | 10 | 1766 | |||
XM_005248542.3 | XP_005248599.1 | 16 | 2957 | |||
XM_011543493.2 | XP_011541795.1 | 15 | 2587 | |||
XM_017009585.1 | XP_016865074.1 | 16 | 3726 | |||
XM_017009586.1 | XP_016865075.1 | 15 | 2547 | |||
GenBank mRNA | AF279367.1 | - | 10 | 1766 | ||
AK094785.1 | - | 9 | 1497 | |||
AK126345.1 | - | 9 | 2897 | |||
AK223613.1 | - | 10 | 1614 | |||
AK299743.1 | - | 14 | 1963 | |||
BC030815.1 | - | 10 | 1606 | 4290954 | ||
BC094795.1 | - | 16 | 2914 | 30528412 | ||
M61906.1 | - | 15 | 2396 |