Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC191 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020817.1 | NP_065868.1 | 4 | 485 | ||
XM_006713715.1 | XP_006713778.1 | 3 | 427 | |||
XM_011513051.1 | XP_011511353.1 | 3 | 934 | |||
XM_011513052.2 | XP_011511354.1 | 2 | 241 | |||
XM_011513053.1 | XP_011511355.1 | 3 | 377 | |||
XM_011513054.2 | XP_011511356.1 | 3 | 213 | |||
XM_017006956.1 | XP_016862445.1 | 2 | 347 | |||
XM_017006957.1 | XP_016862446.1 | 3 | 1032 | |||
XM_017006958.1 | XP_016862447.1 | 3 | 879 | |||
XM_017006959.1 | XP_016862448.1 | 1 | 32 | |||
GenBank mRNA | AF509494.1 | - | 4 | 485 | ||
AK125304.1 | - | 4 | 389 | |||
AK295855.1 | - | 2 | 230 | |||
AK302488.1 | - | 3 | 377 | |||
BC150592.1 | - | 3 | 427 |