Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OSBPL8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003712.1 | NP_001003712.1 | 3, 4 | 658 | ||
NM_001319652.1 | NP_001306581.1 | 6, 7 | 783 | |||
NM_001319653.1 | NP_001306582.1 | 4, 5 | 695 | |||
NM_020841.4 | NP_065892.1 | 4, 5 | 704 | |||
XM_005268621.4 | XP_005268678.1 | 7, 8 | 802 | |||
XM_006719224.2 | XP_006719287.1 | 3, 4 | 663 | |||
XM_011537855.2 | XP_011536157.1 | 4, 5 | 321 | |||
XM_011537856.2 | XP_011536158.1 | 5, 6 | 739 | |||
XM_011537857.2 | XP_011536159.1 | 7, 8 | 793 | |||
XM_017018768.1 | XP_016874257.1 | 4, 5 | 336 | |||
XM_017018769.1 | XP_016874258.1 | 5, 6 | 707 | |||
GenBank mRNA | AB040884.3 | - | 4, 5 | 375 | ||
AF323730.1 | - | 3, 4 | 658 | |||
AF392452.1 | - | 4, 5 | 705 | |||
AK055835.1 | - | 3, 4 | 256 | |||
AK289997.1 | - | 4, 5 | 424 | |||
BC082235.1 | - | 4, 5 | 643 | |||
BC093834.1 | - | 3, 4 | 303 | 7939679 | ||
BC101529.1 | - | 3, 4 | 303 | 8069035 | ||
BC111728.1 | - | 4, 5 | 598 | 40029118 | ||
BX647211.1 | - | 3, 4 | 257 | |||
CR749542.1 | - | 6, 7 | 509 |