Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SCAPER SIRNA Assays ›
Gene Symbol:
SCAPER
Gene Name:
S-phase cyclin A associated protein in the ER
Gene Aliases:
MSTP063, ZNF291, Zfp291
Chromosome Location:
Chr.15: 76348185 - 76905455 on Build GRCh38
UniGene ID:
Hs.458986
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
28885
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SCAPER
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001145923.1
NP_001139395.1
2
156
NM_020843.2
NP_065894.2
3
239
XM_005254417.2
XP_005254474.1
4
396
XM_005254419.2
XP_005254476.1
4
396
XM_011521650.1
XP_011519952.1
5
481
XM_011521652.2
XP_011519954.1
3
192
XM_011521653.2
XP_011519955.1
3
276
XM_017022266.1
XP_016877755.1
4
401
XM_017022267.1
XP_016877756.1
4
401
XM_017022268.1
XP_016877757.1
4
344
XM_017022269.1
XP_016877758.1
4
394
XM_017022270.1
XP_016877759.1
5
552
XM_017022271.1
XP_016877760.1
4
402
XM_017022272.1
XP_016877761.1
4
396
XM_017022273.1
XP_016877762.1
3
276
XM_017022274.1
XP_016877763.1
4
401
XM_017022275.1
XP_016877764.1
4
394
XM_017022276.1
XP_016877765.1
1
49
XM_017022277.1
XP_016877766.1
4
394
XM_017022278.1
XP_016877767.1
3
276
XM_017022279.1
XP_016877768.1
4
398
XM_017022280.1
XP_016877769.1
4
398
XM_017022281.1
XP_016877770.1
4
398
XM_017022282.1
XP_016877771.1
4
397
XM_017022283.1
XP_016877772.1
4
396
XM_017022284.1
XP_016877773.1
4
409
GenBank mRNA
AB040887.1
-
3
226
AF242528.1
-
3
214
AK056095.1
-
4
385
AK056515.1
-
5
456
BC070197.1
-
4
256
BC107415.1
-
4
280
BX647285.1
-
2
156
Back To Top
More Information
Gene Ontology Categories:
Process(es)
nucleic acid binding
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-46vz9:80/10.239.163.144:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0