Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRCH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243963.1 | NP_001230892.1 | 14 | 1700 | ||
NM_020871.3 | NP_065922.3 | 14 | 1700 | |||
XM_006724724.3 | XP_006724787.2 | 14 | 1682 | |||
XM_017029696.1 | XP_016885185.1 | 14 | 1709 | |||
XM_017029697.1 | XP_016885186.1 | 14 | 1710 | |||
GenBank mRNA | AB040928.1 | - | 10 | 881 | ||
AK022128.1 | - | 6 | 456 | |||
BC125223.1 | - | 2 | 107 | |||
BC125224.1 | - | 14 | 1693 | 40124466 | ||
BX647193.1 | - | 14 | 1700 |