Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PMS1 SIRNA Assays ›
Gene Symbol:
PMS1
Gene Name:
PMS1 homolog 1, mismatch repair system component
Gene Aliases:
HNPCC3, MLH2, PMSL1, hPMS1
Chromosome Location:
Chr.2: 189784085 - 189877629 on Build GRCh38
UniGene ID:
Hs.111749
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
2891
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PMS1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000534.4
NP_000525.1
3
723
NM_001128143.1
NP_001121615.1
3
723
NM_001128144.1
NP_001121616.1
3
723
NM_001289408.1
NP_001276337.1
3
723
NM_001321044.1
NP_001307973.1
3
723
NM_001321045.1
NP_001307974.1
4
849
NM_001321047.1
NP_001307976.1
3
900
NM_001321048.1
NP_001307977.1
4
820
NM_001321049.1
NP_001307978.1
3
723
NM_001321051.1
NP_001307980.1
3
723
NR_110332.1
4
820
XM_011511356.2
XP_011509658.1
2
214
XM_017004344.1
XP_016859833.1
4
449
XM_017004348.1
XP_016859837.1
3
449
XM_017004349.1
XP_016859838.1
2
214
XM_017004351.1
XP_016859840.1
2
214
GenBank mRNA
AB102869.1
-
2
194
AB102870.1
-
2
194
AB102873.1
-
2
194
AB102874.1
-
2
194
AB102875.1
-
2
194
AB102876.1
-
2
194
AB102877.1
-
2
194
AK295602.1
-
3
319
AK297440.1
-
3
280
AK308311.1
-
3
319
AY540751.1
-
4
392
BC036376.1
-
3
432
BC084548.1
-
3
300
BC096330.1
-
3
264
BC096331.1
-
3
263
40008290
BC096332.1
-
3
264
BG772717.1
-
3
447
BT006947.1
-
2
194
DA206770.1
-
4
416
DA389799.1
-
3
310
DB178424.1
-
4
447
Back To Top
More Information
Panther Classification:
Molecular Function -
DNA metabolism protein
Gene Ontology Categories:
Function(s)
Process(es)
mismatch repair
DNA binding
single-stranded DNA binding
ATP binding
ATPase activity
mismatched DNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-l9dfq:80/10.239.161.188:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0