Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: TPI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK298809.1 | - | 1470 | |||
BC007812.2 | - | 1207 | 4298901 | |||
BC009329.2 | - | 1213 | 4126160 | |||
BC011611.2 | - | 1212 | 3956298 | |||
BC015100.1 | - | 1227 | 3829453 | |||
BC018899.1 | - | 1637 | ||||
EU794668.1 | - | 1227 |