Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP2A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000762.5 | NP_000753.3 | 2, 3 | 364 | ||
GenBank mRNA | AF182275.1 | - | 2, 3 | 352 | ||
AK307740.1 | - | 2, 3 | 374 | |||
AK312964.1 | - | 2, 3 | 364 | |||
BC096253.3 | - | 2, 3 | 352 | 40006061 | ||
BC096254.1 | - | 2, 3 | 352 | 40006062 | ||
BC096255.1 | - | 2, 3 | 352 | 40006064 | ||
BC096256.1 | - | 2, 3 | 352 | 40006068 | ||
DA641580.1 | - | 2, 3 | 364 | |||
M33318.1 | - | 2, 3 | 352 | |||
X13897.1 | - | 2, 3 | 338 | |||
X13929.1 | - | 2, 3 | 206 | |||
X13930.1 | - | 2, 3 | 352 |