Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTGER1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000955.2 | NP_000946.2 | 1418 | |||
GenBank mRNA | BC039034.1 | - | 1367 | 5748278 | ||
BC051286.1 | - | 1418 | 6596880 |
Set Membership: |
G-protein coupled receptor |