Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AFMID | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010982.4 | NP_001010982.2 | 10 | 855 | ||
NM_001145526.2 | NP_001138998.1 | 10 | 870 | |||
XM_011524326.2 | XP_011522628.1 | 9 | 802 | |||
XM_011524327.2 | XP_011522629.1 | 11 | 1038 | |||
XM_011524328.2 | XP_011522630.1 | 11 | 1010 | |||
XM_011524329.2 | XP_011522631.1 | 11 | 1052 | |||
XM_011524330.2 | XP_011522632.1 | 11 | 1056 | |||
XM_011524331.2 | XP_011522633.1 | 10 | 952 | |||
XM_011524332.2 | XP_011522634.1 | 10 | 924 | |||
XM_011524333.2 | XP_011522635.1 | 9 | 678 | |||
XM_011524334.2 | XP_011522636.1 | 8 | 781 | |||
XM_011524335.2 | XP_011522637.1 | 8 | 766 | |||
XM_011524336.2 | XP_011522638.1 | 7 | 501 | |||
XM_011524337.2 | XP_011522639.1 | 6 | 457 | |||
GenBank mRNA | BC132824.1 | - | 10 | 855 | 40146846 | |
BX648442.1 | - | 10 | 826 |