Gene Symbol: GCNT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001491.2 | NP_001482.1 | 3 | 2041 | ||
NM_145649.4 | NP_663624.1 | 5 | 1890 | |||
NM_145655.3 | NP_663630.2 | 3 | 1569 | |||
XM_005248999.2 | XP_005249056.1 | 6 | 1688 | |||
XM_006715052.3 | XP_006715115.1 | 5 | 1967 | |||
GenBank mRNA | AK090483.1 | - | 3 | 1895 | ||
AK098105.1 | - | 3 | 733 | |||
AK291195.1 | - | 3 | 1625 | |||
AK291767.1 | - | 5 | 1874 | |||
AK307329.1 | - | 4 | 697 | |||
AK307337.1 | - | 5 | 770 | |||
AK307373.1 | - | 5 | 898 | |||
AL832714.1 | - | 3 | 592 | |||
AL832719.1 | - | 3 | 1570 | |||
AY435145.1 | - | 3 | 1339 | |||
AY435146.1 | - | 3 | 1333 | |||
AY435147.1 | - | 3 | 1339 | |||
BC130524.1 | - | 3 | 1511 | 40146555 | ||
BX647576.1 | - | 3 | 1568 | |||
BX647668.1 | - | 3 | 1570 | |||
L19659.1 | - | 3 | 1587 | |||
Z19550.1 | - | 3 | 1587 |