Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CST1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001898.2 | NP_001889.2 | 1 | 96 | ||
GenBank mRNA | BC021225.2 | - | 1 | 94 | 4298603 | |
J03870.1 | - | 1 | 95 |