Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: MIF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002415.1 | NP_002406.1 | 3 | 549 | ||
GenBank mRNA | BC000447.2 | - | 3 | 533 | 2821346 | |
BC007676.1 | - | 3 | 517 | 3605756 | ||
BC008914.2 | - | 3 | 503 | 3615896 | ||
BC013976.2 | - | 3 | 548 | 4130651 | ||
BC022414.1 | - | 3 | 550 | 4243461 | ||
BC053376.1 | - | 3 | 486 | 6047427 | ||
L10612.1 | - | 3 | 526 | |||
M25639.1 | - | 3 | 502 | |||
M95775.1 | - | 3 | 426 |